missing translation for 'onlineSavingsMsg'
Learn More

(4-Methoxyphenyl)urea 98.0+%, TCI America™

Catalog No. M25751G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25751G 1 g
M25755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25751G Supplier TCI America Supplier No. M25751G

Chemical Identifiers

CAS 1566-42-3
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00014789
InChI Key PGUKYDVWVXRPKK-UHFFFAOYSA-N
Synonym 4-methoxyphenyl urea, urea, 4-methoxyphenyl, p-anisylurea, 1-4-methoxyphenyl urea, urea, p-methoxyphenyl, p-methoxyphenyl urea, p-methoxyphenylurea, n-4-methoxyphenyl urea, amino-n-4-methoxyphenyl amide, acmc-20amej
PubChem CID 74066
IUPAC Name (4-methoxyphenyl)urea
SMILES COC1=CC=C(NC(N)=O)C=C1

Specifications

Melting Point 162°C
Color White
Quantity 1 g
Formula Weight 166.18
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (4-Methoxyphenyl)urea
RTECSNumber YT6451200
TSCA No