missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenyl Trifluoromethanesulfonate 97.0+%, TCI America™

Catalog No. M20445G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M20445G Supplier TCI America Supplier No. M20445G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 66107-29-7
Molecular Formula C8H7F3O4S
Molecular Weight (g/mol) 256.195
MDL Number MFCD00209596
InChI Key XSTNIRWDULKNJE-UHFFFAOYSA-N
Synonym Trifluoromethanesulfonic Acid 4-Methoxyphenyl Ester, 4-Methoxyphenyl Triflate
PubChem CID 579337
IUPAC Name (4-methoxyphenyl) trifluoromethanesulfonate
SMILES COC1=CC=C(C=C1)OS(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 92°C
Quantity 5 g
Formula Weight 256.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methoxyphenyl Trifluoromethanesulfonate