accessibility menu, dialog, popup

UserName

CAS RN 66107-29-7

IUPAC Name: 4-methoxyphenyl trifluoromethanesulfonate
Synonyms: 4-methoxyphenyl trifluoromethanesulfonate
Molecular Weight (g/mol): 256.2
Molecular Formula: C8H7F3O4S
InChi Key: XSTNIRWDULKNJE-UHFFFAOYSA-N
SMILES: COC1=CC=C(OS(=O)(=O)C(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results