missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenyl trans-4-n-pentylcyclohexanecarboxylate, 97%, Thermo Scientific™

Catalog No. p-7037161
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5579203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5579203 Supplier Thermo Scientific Chemicals Supplier No. H5579203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Methoxyphenyl trans-4-n-pentylcyclohexanecarboxylate, is mainly used as a nematic liquid crystal monomer, liquid crystal material. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Sparingly Soluble in water (1.7E-3 g/L) (25°C).

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 67589-52-0
Molecular Formula C19H28O3
Molecular Weight (g/mol) 304.43
MDL Number MFCD11053432
InChI Key HJIVFZUPTYADSP-UHFFFAOYSA-N
Synonym cyclohexanecarboxylic acid, 4-pentyl-, 4-methoxyphenyl ester, trans, trans-4-methoxyphenyl 4-pentylcyclohexanecarboxylate, trans-4-methoxy-phenyl 4-pentyl-cyclohexanecarboxylate, trans-4-pentylcyclohexanecarboxylic acid, 4-methoxyphenyl ester, 4-methoxyphenyl trans-4-pentylcyclohexanoate, 4-methoxyphenyl 4-pentylcyclohexanecarboxylate, 4-methoxyphenyl trans-4-pentylcyclohexanecarboxylate, hjivfzuptyadsp-wkilwmfisa-n, 4-methoxyphenyl 4-pentylcyclohexanecarboxylate #, 4-methoxyphenyl 4-pentylcyclohexane-1-carboxylate
PubChem CID 105460
IUPAC Name (4-methoxyphenyl) 4-pentylcyclohexane-1-carboxylate
SMILES CCCCCC1CCC(CC1)C(=O)OC2=CC=C(C=C2)OC

Specifications

Melting Point 40°C to 44°C
Quantity 1 g
Solubility Information Sparingly Soluble in water (1.7E-3g/L) (25°C).
Formula Weight 304.43
Percent Purity 97%
Chemical Name or Material 4-Methoxyphenyl trans-4-n-pentylcyclohexanecarboxylate
EINECSNumber 266-746-6
TSCA Yes
Recommended Storage Ambient temperatures