missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxyphenyl Isocyanate 98.0+%, TCI America™
Supplier: TCI America I044025G
Specifications
| 4-Methoxyphenyl Isocyanate | |
| Yellow | |
| C8H7NO2 | |
| 25 g | |
| 4-methoxyphenyl isocyanate, 4-methoxyphenylisocyanate, benzene, 1-isocyanato-4-methoxy, p-anisyl isocyanate, p-methoxyphenyl isocyanate, isocyanic acid, p-methoxyphenyl ester, unii-c50o5096nc, 1-isocyanato-4-methoxy-benzene, 4-methoxybenzenisocyanate, 4-methoxyisocyanatobenzene | |
| COC1=CC=C(C=C1)N=C=O | |
| 149.149 | |
| 149.15 | |
| Liquid |
| 5416-93-3 | |
| 133°C | |
| MFCD00002026 | |
| 2206 | |
| FMDGXCSMDZMDHZ-UHFFFAOYSA-N | |
| 1-isocyanato-4-methoxybenzene | |
| 79443 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 5416-93-3 | |
| 149.149 | |
| FMDGXCSMDZMDHZ-UHFFFAOYSA-N | |
| 79443 | |
| COC1=CC=C(C=C1)N=C=O |
| C8H7NO2 | |
| MFCD00002026 | |
| 4-methoxyphenyl isocyanate, 4-methoxyphenylisocyanate, benzene, 1-isocyanato-4-methoxy, p-anisyl isocyanate, p-methoxyphenyl isocyanate, isocyanic acid, p-methoxyphenyl ester, unii-c50o5096nc, 1-isocyanato-4-methoxy-benzene, 4-methoxybenzenisocyanate, 4-methoxyisocyanatobenzene | |
| 1-isocyanato-4-methoxybenzene |
Safety and Handling
TSCA : Yes