missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenyl isocyanate, 99%

CAS: 5416-93-3 | C8H7NO2 | 149.15 g/mol

$328.21 - $328.21

Chemical Identifiers

CAS 5416-93-3
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.15
MDL Number MFCD00002026
InChI Key FMDGXCSMDZMDHZ-UHFFFAOYSA-N
Synonym 4-methoxyphenyl isocyanate, 4-methoxyphenylisocyanate, benzene, 1-isocyanato-4-methoxy, p-anisyl isocyanate, p-methoxyphenyl isocyanate, isocyanic acid, p-methoxyphenyl ester, unii-c50o5096nc, 1-isocyanato-4-methoxy-benzene, 4-methoxybenzenisocyanate, 4-methoxyisocyanatobenzene
PubChem CID 79443
IUPAC Name 1-isocyanato-4-methoxybenzene
SMILES COC1=CC=C(C=C1)N=C=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC218760250
SDS View Documents
Thermo Scientific Chemicals
218760250
25 g Glass bottle
Each for $328.21
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5416-93-3
149.15
FMDGXCSMDZMDHZ-UHFFFAOYSA-N
79443
COC1=CC=C(C=C1)N=C=O
C8H7NO2
MFCD00002026
4-methoxyphenyl isocyanate, 4-methoxyphenylisocyanate, benzene, 1-isocyanato-4-methoxy, p-anisyl isocyanate, p-methoxyphenyl isocyanate, isocyanic acid, p-methoxyphenyl ester, unii-c50o5096nc, 1-isocyanato-4-methoxy-benzene, 4-methoxybenzenisocyanate, 4-methoxyisocyanatobenzene
1-isocyanato-4-methoxybenzene

Specifications

5416-93-3
100.0
1.1500g/mL
98°C
98.5% min. (GC)
C8H7NO2
CH3OC6H4NCO
25 g
1.15
FMDGXCSMDZMDHZ-UHFFFAOYSA-N
1-isocyanato-4-methoxybenzene
79443
99%
4-Methoxyphenyl isocyanate, 99%
98.5
Colorless
106°C to 110°C (16.0 mmHg)
Authentic
Glass bottle
1.5450 to 1.547
MFCD00002026
13, 487
4-methoxyphenyl isocyanate, 4-methoxyphenylisocyanate, benzene, 1-isocyanato-4-methoxy, p-anisyl isocyanate, p-methoxyphenyl isocyanate, isocyanic acid, p-methoxyphenyl ester, unii-c50o5096nc, 1-isocyanato-4-methoxy-benzene, 4-methoxybenzenisocyanate, 4-methoxyisocyanatobenzene
COC1=CC=C(C=C1)N=C=O
149.15
149.15
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 226-513-1

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only