missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenyl isocyanate, 99%

Catalog No. AC218760050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC218760050 5 g Glass bottle
AC218760250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC218760050 Supplier Thermo Scientific Chemicals Supplier No. 218760050

CAS: 5416-93-3 | C8H7NO2 | 149.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5416-93-3
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.15
MDL Number MFCD00002026
InChI Key FMDGXCSMDZMDHZ-UHFFFAOYSA-N
Synonym 4-methoxyphenyl isocyanate, 4-methoxyphenylisocyanate, benzene, 1-isocyanato-4-methoxy, p-anisyl isocyanate, p-methoxyphenyl isocyanate, isocyanic acid, p-methoxyphenyl ester, unii-c50o5096nc, 1-isocyanato-4-methoxy-benzene, 4-methoxybenzenisocyanate, 4-methoxyisocyanatobenzene
PubChem CID 79443
IUPAC Name 1-isocyanato-4-methoxybenzene
SMILES COC1=CC=C(C=C1)N=C=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 1.1500g/mL
Boiling Point 106°C to 110°C (16.0 mmHg)
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5450 to 1.547
Linear Formula CH3OC6H4NCO
Quantity 5 g
Beilstein 13, 487
Specific Gravity 1.15
Formula Weight 149.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Methoxyphenyl isocyanate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 226-513-1
TSCA TSCA

RUO – Research Use Only