missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenyl isocyanate, 98%

Catalog No. AAA1680506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1680506 5 g
AAA1680514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1680506 Supplier Thermo Scientific Chemicals Supplier No. A1680506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5416-93-3 | C8H7NO2 | 149.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5416-93-3
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.149
MDL Number MFCD00002026
InChI Key FMDGXCSMDZMDHZ-UHFFFAOYSA-N
Synonym 4-methoxyphenyl isocyanate, 4-methoxyphenylisocyanate, benzene, 1-isocyanato-4-methoxy, p-anisyl isocyanate, p-methoxyphenyl isocyanate, isocyanic acid, p-methoxyphenyl ester, unii-c50o5096nc, 1-isocyanato-4-methoxy-benzene, 4-methoxybenzenisocyanate, 4-methoxyisocyanatobenzene
PubChem CID 79443
IUPAC Name 1-isocyanato-4-methoxybenzene
SMILES COC1=CC=C(C=C1)N=C=O

Specifications

Density 1.151
Boiling Point 119°C to 120°C (20 mmHg)
Flash Point 98°C (208°F)
Odor Pungent
Refractive Index 1.547
Quantity 5 g
UN Number UN2206
Beilstein 471920
Sensitivity Moisture sensitive
Formula Weight 149.15
Percent Purity 98%
Chemical Name or Material 4-Methoxyphenyl isocyanate
Show More Show Less
Hazard Category H302-H312-H314-H332-H335
Hazard Statement GHS H Statement
H334-H302-H332-H335-H315-H319
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 226-513-1
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only