missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxyphenyl alpha-D-Mannopyranoside 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 156°C |
| Quantity | 5 g |
| Formula Weight | 286.28 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Methoxyphenyl alpha-D-Mannopyranoside |
Safety and Handling
| TSCA | No |
| Recommended Storage | Freezer |