accessibility menu, dialog, popup

UserName

H3COOOOHOHOHOH

CAS RN 28541-75-5

IUPAC Name: 2-(hydroxymethyl)-6-(4-methoxyphenoxy)oxane-3,4,5-triol
Synonyms: 2-(hydroxymethyl)-6-(4-methoxyphenoxy)oxane-3,4,5-triol
Molecular Weight (g/mol): 286.28
Molecular Formula: C13H18O7
InChi Key: SIXFVXJMCGPTRB-UHFFFAOYNA-N
SMILES: COC1=CC=C(OC2OC(CO)C(O)C(O)C2O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results