missing translation for 'onlineSavingsMsg'
Learn More

(4-Methoxyphenyl)acetonitrile, 97%

Catalog No. p-3758316
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC173760250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC173760250 Supplier Thermo Scientific Chemicals Supplier No. 173760250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 104-47-2 | C9H9NO | 147.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104-47-2
MDL Number MFCD00001919
InChI Key PACGLQCRGWFBJH-UHFFFAOYSA-N
Synonym 4-methoxyphenylacetonitrile, 2-4-methoxyphenyl acetonitrile, 4-methoxybenzyl cyanide, 4-methoxyphenyl acetonitrile, p-methoxyphenylacetonitrile, benzeneacetonitrile, 4-methoxy, p-methoxybenzyl cyanide, anisylacetonitrile, 4-anisyl cyanide, p-methoxybenzeneacetonitrile
PubChem CID 66031
IUPAC Name 2-(4-methoxyphenyl)acetonitrile
SMILES COC1=CC=C(C=C1)CC#N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 8.0°C
Color Colorless to Yellow
Density 1.0800g/mL
Boiling Point 286.0°C to 287.0°C
Flash Point 117°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C9H9NO
Refractive Index 1.53
Linear Formula CH3OC6H4CH2CN
Quantity 25 g
Beilstein 10, 191
Specific Gravity 1.08
Solubility Information Solubility in water: 10g/L (20°C). Other solubilities: soluble in ethyl alcohol,ethyl ether,chloroform,miscible in isopropanol,acetone,toluene
Molecular Weight (g/mol) 147.18
Formula Weight 147.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (4-Methoxyphenyl)acetonitrile
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Rinse mouth.
Avoid r
EINECSNumber 203-206-

RUO – Research Use Only