missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenyl 4-O-Acetyl-3,6-di-O-benzyl-2-deoxy-2-phthalimido-beta-D-glucopyranoside 98.0+%, TCI America™

Supplier:  TCI America M18345G

 View more versions of this product

Catalog No. M18345G


Only null left

Description

Description

Specifications

Specifications

4-Methoxyphenyl 4-O-Acetyl-3,6-di-O-benzyl-2-deoxy-2-phthalimido-beta-D-glucopyranoside
White
5 g
QZTMIUALKXAHCQ-RHPHZSOCSA-N
[(2R,3R,4S,5R,6R)-5-(1,3-dioxoisoindol-2-yl)-6-(4-methoxyphenoxy)-4-phenylmethoxy-2-(phenylmethoxymethyl)oxan-3-yl] acetate
133554185
≥98.0% (HPLC)
140615-77-6
C37H35NO9
GlcNPhth[36Bn,4Ac]-beta-MP
CC(=O)OC1C(OC(C(C1OCC2=CC=CC=C2)N3C(=O)C4=CC=CC=C4C3=O)OC5=CC=C(C=C5)OC)COCC6=CC=CC=C6
637.685
637.69
Crystalline Powder

Chemical Identifiers

140615-77-6
637.685
GlcNPhth[36Bn,4Ac]-beta-MP
[(2R,3R,4S,5R,6R)-5-(1,3-dioxoisoindol-2-yl)-6-(4-methoxyphenoxy)-4-phenylmethoxy-2-(phenylmethoxymethyl)oxan-3-yl] acetate
C37H35NO9
QZTMIUALKXAHCQ-RHPHZSOCSA-N
133554185
CC(=O)OC1C(OC(C(C1OCC2=CC=CC=C2)N3C(=O)C4=CC=CC=C4C3=O)OC5=CC=C(C=C5)OC)COCC6=CC=CC=C6
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions