accessibility menu, dialog, popup

UserName

CAS RN 140615-77-6

IUPAC Name: 4-(benzyloxy)-2-[(benzyloxy)methyl]-5-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)-6-(4-methoxyphenoxy)oxan-3-yl acetate
Synonyms: 4-(benzyloxy)-2-[(benzyloxy)methyl]-5-(1,3-dioxoisoindol-2-yl)-6-(4-methoxyphenoxy)oxan-3-yl acetate
Molecular Weight (g/mol): 637.69
Molecular Formula: C37H35NO9
InChi Key: QZTMIUALKXAHCQ-UHFFFAOYNA-N
SMILES: COC1=CC=C(OC2OC(COCC3=CC=CC=C3)C(OC(C)=O)C(OCC3=CC=CC=C3)C2N2C(=O)C3=CC=CC=C3C2=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results