missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenyl 4-(3-Butenyloxy)benzoate 98.0+%, TCI America™

Catalog No. M21061G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21061G 1 g
M21065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21061G Supplier TCI America Supplier No. M21061G

Chemical Identifiers

CAS 76487-56-4
Molecular Formula C18H18O4
Molecular Weight (g/mol) 298.34
MDL Number MFCD00143337
InChI Key CXPLNUIZUUBDCU-UHFFFAOYSA-N
Synonym 4-(3-Butenyloxy)benzoic Acid 4-Methoxyphenyl Ester
PubChem CID 144778
IUPAC Name 4-methoxyphenyl 4-(but-3-en-1-yloxy)benzoate
SMILES COC1=CC=C(OC(=O)C2=CC=C(OCCC=C)C=C2)C=C1

Specifications

Color White
Quantity 1 g
Formula Weight 298.34
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenyl 4-(3-Butenyloxy)benzoate