accessibility menu, dialog, popup

UserName

CAS RN 76487-56-4

IUPAC Name: 4-methoxyphenyl 4-(but-3-en-1-yloxy)benzoate
Synonyms: 4-methoxyphenyl 4-(but-3-en-1-yloxy)benzoate
Molecular Weight (g/mol): 298.34
Molecular Formula: C18H18O4
InChi Key: CXPLNUIZUUBDCU-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC(=O)C2=CC=C(OCCC=C)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results