missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenyl 3,4-O-Isopropylidene-beta-D-galactopyranoside 98.0+%, TCI America™

Catalog No. M15935G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M15935G 5 g
M15931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M15935G Supplier TCI America Supplier No. M15935G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 159922-67-5 | C16H22O7 | 326.345 g/mol

Chemical Identifiers

CAS 159922-67-5
Molecular Formula C16H22O7
Molecular Weight (g/mol) 326.345
MDL Number MFCD06797135
InChI Key IFIVVNHQGPJZBP-UXXRCYHCSA-N
PubChem CID 44629762
IUPAC Name (3aS,4R,6S,7R,7aR)-4-(hydroxymethyl)-6-(4-methoxyphenoxy)-2,2-dimethyl-4,6,7,7a-tetrahydro-3aH-[1,3]dioxolo[4,5-c]pyran-7-ol
SMILES CC1(OC2C(OC(C(C2O1)O)OC3=CC=C(C=C3)OC)CO)C

Specifications

Melting Point 151°C
Color White
Quantity 5 g
Formula Weight 326.35
Percent Purity ≥98.0% (HPLC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenyl 3,4-O-Isopropylidene-beta-D-galactopyranoside
TSCA No
Recommended Storage Freezer