accessibility menu, dialog, popup

UserName

H3COOOOHOOCH3OHH3C

CAS RN 159922-67-5

IUPAC Name: 4-(hydroxymethyl)-6-(4-methoxyphenoxy)-2,2-dimethyl-hexahydro-[1,3]dioxolo[4,5-c]pyran-7-ol
Synonyms: 4-(hydroxymethyl)-6-(4-methoxyphenoxy)-2,2-dimethyl-tetrahydro-3aH-[1,3]dioxolo[4,5-c]pyran-7-ol
Molecular Weight (g/mol): 326.35
Molecular Formula: C16H22O7
InChi Key: IFIVVNHQGPJZBP-UHFFFAOYNA-N
SMILES: COC1=CC=C(OC2OC(CO)C3OC(C)(C)OC3C2O)C=C1
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)