missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxyphenyl 3,4-O-Isopropylidene-beta-D-galactopyranoside 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 151°C |
| Color | White |
| Quantity | 1 g |
| Formula Weight | 326.35 |
| Percent Purity | ≥98.0% (HPLC) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Methoxyphenyl 3,4-O-Isopropylidene-beta-D-galactopyranoside |
Safety and Handling
| TSCA | No |
| Recommended Storage | Freezer |