missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenyl 2-Azido-4,6-O-benzylidene-2-deoxy-beta-D-glucopyranoside 98.0+%, TCI America™

Catalog No. m16371g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16371G 1 g
M16375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M16371G Supplier TCI America Supplier No. M16371G

Chemical Identifiers

CAS 1430068-18-0
Molecular Formula C20H21N3O6
Molecular Weight (g/mol) 399.403
MDL Number MFCD06797146
InChI Key HCOPARKYIRRSBS-BZIXAJQCSA-N
PubChem CID 44629750
IUPAC Name (2R,4aR,6S,7R,8R,8aS)-7-azido-6-(4-methoxyphenoxy)-2-phenyl-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-8-ol
SMILES COC1=CC=C(C=C1)OC2C(C(C3C(O2)COC(O3)C4=CC=CC=C4)O)N=[N+]=[N-]

Specifications

Melting Point 165°C
Color White
Quantity 1 g
Formula Weight 399.40
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenyl 2-Azido-4,6-O-benzylidene-2-deoxy-beta-D-glucopyranoside
TSCA No
Recommended Storage Freezer