accessibility menu, dialog, popup

UserName

CAS RN 1430068-18-0

IUPAC Name: 7-azido-6-(4-methoxyphenoxy)-2-phenyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-8-ol
Synonyms: 7-azido-6-(4-methoxyphenoxy)-2-phenyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-8-ol
Molecular Weight (g/mol): 399.4
Molecular Formula: C20H21N3O6
InChi Key: HCOPARKYIRRSBS-UHFFFAOYNA-N
SMILES: COC1=CC=C(OC2OC3COC(OC3C(O)C2N=[N+]=[N-])C2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results