missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenyl 2-Amino-3,6-di-O-benzyl-2-deoxy-beta-D-glucopyranoside, TCI America™

Catalog No. M16161G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M16161G Supplier TCI America Supplier No. M16161G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1272755-07-3
Molecular Formula C27H31NO6
Molecular Weight (g/mol) 465.546
InChI Key DFPWFVXGRBQWGQ-RFNQJFSXSA-N
PubChem CID 44629780
IUPAC Name (2R,3S,4R,5R,6S)-5-amino-6-(4-methoxyphenoxy)-4-phenylmethoxy-2-(phenylmethoxymethyl)oxan-3-ol
SMILES COC1=CC=C(C=C1)OC2C(C(C(C(O2)COCC3=CC=CC=C3)O)OCC4=CC=CC=C4)N

Specifications

Melting Point 105°C
Color White-Yellow
Quantity 1 g
Formula Weight 465.55
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenyl 2-Amino-3,6-di-O-benzyl-2-deoxy-beta-D-glucopyranoside
TSCA No
Recommended Storage Freezer