missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxyphenyl 2,6-Di-O-benzyl-beta-D-galactopyranoside 98.0+%, TCI America™
$82.02 - $244.19
Chemical Identifiers
| CAS | 159922-50-6 |
|---|---|
| Molecular Formula | C27H30O7 |
| Molecular Weight (g/mol) | 466.53 |
| MDL Number | MFCD06797145 |
| InChI Key | ZNONEMGWLHXQDN-LXSUACKSSA-N |
| PubChem CID | 10906733 |
| IUPAC Name | (2R,3R,4S,5R,6S)-6-(4-methoxyphenoxy)-5-phenylmethoxy-2-(phenylmethoxymethyl)oxane-3,4-diol |
| SMILES | COC1=CC=C(C=C1)OC2C(C(C(C(O2)COCC3=CC=CC=C3)O)O)OCC4=CC=CC=C4 |
Chemical Identifiers
| 159922-50-6 | |
| 466.53 | |
| ZNONEMGWLHXQDN-LXSUACKSSA-N | |
| (2R,3R,4S,5R,6S)-6-(4-methoxyphenoxy)-5-phenylmethoxy-2-(phenylmethoxymethyl)oxane-3,4-diol |
| C27H30O7 | |
| MFCD06797145 | |
| 10906733 | |
| COC1=CC=C(C=C1)OC2C(C(C(C(O2)COCC3=CC=CC=C3)O)O)OCC4=CC=CC=C4 |
Specifications
| 159922-50-6 | |
| White-Yellow | |
| MFCD06797145 | |
| ZNONEMGWLHXQDN-LXSUACKSSA-N | |
| (2R,3R,4S,5R,6S)-6-(4-methoxyphenoxy)-5-phenylmethoxy-2-(phenylmethoxymethyl)oxane-3,4-diol | |
| 10906733 | |
| ≥98.0% (HPLC) | |
| 4-Methoxyphenyl 2,6-Di-O-benzyl-beta-D-galactopyranoside |
| 113°C | |
| C27H30O7 | |
| 1 g | |
| COC1=CC=C(C=C1)OC2C(C(C(C(O2)COCC3=CC=CC=C3)O)O)OCC4=CC=CC=C4 | |
| 466.53 | |
| 466.53 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer