accessibility menu, dialog, popup

UserName

CAS RN 159922-50-6

H3COOOOHOOHO

CAS RN 159922-50-6

IUPAC Name: 5-(benzyloxy)-2-[(benzyloxy)methyl]-6-(4-methoxyphenoxy)oxane-3,4-diol
Synonyms: 5-(benzyloxy)-2-[(benzyloxy)methyl]-6-(4-methoxyphenoxy)oxane-3,4-diol
Molecular Weight (g/mol): 466.53
Molecular Formula: C27H30O7
InChi Key: ZNONEMGWLHXQDN-UHFFFAOYNA-N
SMILES: COC1=CC=C(OC2OC(COCC3=CC=CC=C3)C(O)C(O)C2OCC2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results