missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenyl 2,6-Di-O-benzyl-beta-D-galactopyranoside 98.0+%, TCI America™

Catalog No. M16345G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16345G 5 g
M16341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M16345G Supplier TCI America Supplier No. M16345G
Only null left

Chemical Identifiers

CAS 159922-50-6
Molecular Formula C27H30O7
Molecular Weight (g/mol) 466.53
MDL Number MFCD06797145
InChI Key ZNONEMGWLHXQDN-LXSUACKSSA-N
PubChem CID 10906733
IUPAC Name (2R,3R,4S,5R,6S)-6-(4-methoxyphenoxy)-5-phenylmethoxy-2-(phenylmethoxymethyl)oxane-3,4-diol
SMILES COC1=CC=C(C=C1)OC2C(C(C(C(O2)COCC3=CC=CC=C3)O)O)OCC4=CC=CC=C4

Specifications

Melting Point 113°C
Color White-Yellow
Quantity 5 g
Formula Weight 466.53
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenyl 2,6-Di-O-benzyl-beta-D-galactopyranoside
TSCA No
Recommended Storage Freezer