missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxyphenyl 2,6-Di-O-benzyl-3,4-O-isopropylidene-beta-D-galactopyranoside 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 105°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 506.60 |
| Percent Purity | ≥98.0% (HPLC) |
| Grade | Laboratory |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Methoxyphenyl 2,6-Di-O-benzyl-3,4-O-isopropylidene-beta-D-galactopyranoside |
Safety and Handling
| TSCA | No |
| Recommended Storage | Freezer |