CAS RN 159922-68-6
IUPAC Name:
7-(benzyloxy)-4-[(benzyloxy)methyl]-6-(4-methoxyphenoxy)-2,2-dimethyl-hexahydro-[1,3]dioxolo[4,5-c]pyran
Synonyms:
7-(benzyloxy)-4-[(benzyloxy)methyl]-6-(4-methoxyphenoxy)-2,2-dimethyl-tetrahydro-3aH-[1,3]dioxolo[4,5-c]pyran
Molecular Weight (g/mol):
506.6
Molecular Formula:
C30H34O7
InChi Key:
PGZOKVMOUQDWPV-UHFFFAOYNA-N
SMILES:
COC1=CC=C(OC2OC(COCC3=CC=CC=C3)C3OC(C)(C)OC3C2OCC2=CC=CC=C2)C=C1
1
–
1
of
1
results
1
4-Methoxyphenyl 2,6-Di-O-benzyl-3,4-O-isopropylidene-beta-D-galactopyranoside 98.0+%, TCI America™
CAS: 159922-68-6 Molecular Formula: C30H34O7 Molecular Weight (g/mol): 506.595 MDL Number: MFCD06797144 InChI Key: PGZOKVMOUQDWPV-JYJZCUDQSA-N PubChem CID: 24773293 IUPAC Name: (3aS,4R,6S,7R,7aS)-6-(4-methoxyphenoxy)-2,2-dimethyl-7-phenylmethoxy-4-(phenylmethoxymethyl)-4,6,7,7a-tetrahydro-3aH-[1,3]dioxolo[4,5-c]pyran SMILES: CC1(OC2C(OC(C(C2O1)OCC3=CC=CC=C3)OC4=CC=C(C=C4)OC)COCC5=CC=CC=C5)C