missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenyl 2,6-Di-O-benzyl-3,4-O-isopropylidene-beta-D-galactopyranoside 98.0+%, TCI America™

Catalog No. M16331G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16331G 1 g
M16335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M16331G Supplier TCI America Supplier No. M16331G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 159922-68-6 | C30H34O7 | 506.595 g/mol

Chemical Identifiers

CAS 159922-68-6
Molecular Formula C30H34O7
Molecular Weight (g/mol) 506.595
MDL Number MFCD06797144
InChI Key PGZOKVMOUQDWPV-JYJZCUDQSA-N
PubChem CID 24773293
IUPAC Name (3aS,4R,6S,7R,7aS)-6-(4-methoxyphenoxy)-2,2-dimethyl-7-phenylmethoxy-4-(phenylmethoxymethyl)-4,6,7,7a-tetrahydro-3aH-[1,3]dioxolo[4,5-c]pyran
SMILES CC1(OC2C(OC(C(C2O1)OCC3=CC=CC=C3)OC4=CC=C(C=C4)OC)COCC5=CC=CC=C5)C

Specifications

Melting Point 105°C
Color White
Quantity 1 g
Formula Weight 506.60
Percent Purity ≥98.0% (HPLC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenyl 2,6-Di-O-benzyl-3,4-O-isopropylidene-beta-D-galactopyranoside
TSCA No
Recommended Storage Freezer