missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenyl 2,6-Bis-O-(4-methylbenzoyl)-beta-D-galactopyranoside 98.0+%, TCI America™

Catalog No. m15971g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M15971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M15971G Supplier TCI America Supplier No. M15971G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1820570-59-9
Molecular Formula C29H30O9
Molecular Weight (g/mol) 522.55
MDL Number MFCD06797138
InChI Key KGTSZMZIXWBXKC-MXCNSHPNSA-N
PubChem CID 44629752
IUPAC Name [(2R,3R,4S,5R,6S)-3,4-dihydroxy-6-(4-methoxyphenoxy)-5-(4-methylbenzoyl)oxyoxan-2-yl]methyl 4-methylbenzoate
SMILES CC1=CC=C(C=C1)C(=O)OCC2C(C(C(C(O2)OC3=CC=C(C=C3)OC)OC(=O)C4=CC=C(C=C4)C)O)O

Specifications

Melting Point 205°C
Color White
Quantity 1 g
Formula Weight 522.55
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenyl 2,6-Bis-O-(4-methylbenzoyl)-beta-D-galactopyranoside
TSCA No
Recommended Storage Freezer