accessibility menu, dialog, popup

UserName

CAS RN 1820570-59-9

CH3OOOOOCH3OOH3COHOH

CAS RN 1820570-59-9

IUPAC Name: [3,4-dihydroxy-6-(4-methoxyphenoxy)-5-(4-methylbenzoyloxy)oxan-2-yl]methyl 4-methylbenzoate
Synonyms: [3,4-dihydroxy-6-(4-methoxyphenoxy)-5-(4-methylbenzoyloxy)oxan-2-yl]methyl 4-methylbenzoate
Molecular Weight (g/mol): 522.55
Molecular Formula: C29H30O9
InChi Key: KGTSZMZIXWBXKC-UHFFFAOYNA-N
SMILES: COC1=CC=C(OC2OC(COC(=O)C3=CC=C(C)C=C3)C(O)C(O)C2OC(=O)C2=CC=C(C)C=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results