missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenoxyacetic acid, 98%

Catalog No. p-7021895
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1441909 10 g
AAA1441918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1441909 Supplier Thermo Scientific Chemicals Supplier No. A1441909
Only null left

CAS: 1877-75-4 | C9H10O4 | 182.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1877-75-4
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00014360
InChI Key BHFSBJHPPFJCOS-UHFFFAOYSA-N
Synonym 4-methoxyphenoxyacetic acid, 2-4-methoxyphenoxy acetic acid, 4-methoxyphenoxy acetic acid, acetic acid, 4-methoxyphenoxy, p-methoxyphenoxyacetic acid, acetic acid, p-methoxyphenoxy, acetic acid, 2-4-methoxyphenoxy, p-methoxyphenoxy acetic acid, akos bb/0049, rarechem al bo 0321
PubChem CID 74649
IUPAC Name 2-(4-methoxyphenoxy)acetic acid
SMILES COC1=CC=C(OCC(O)=O)C=C1

Specifications

Melting Point 109°C to 113°C
Quantity 10 g
Beilstein 1874579
Formula Weight 182.18
Percent Purity 98%
Chemical Name or Material 4-Methoxyphenoxyacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-513-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only