missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 4-Methoxyphenol

Numéro de catalogue. M186551KG
Encompass
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
M186551KG 1 kg
M18655100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. M186551KG Fournisseur Sigma Aldrich Code fournisseur M186551KG
Il en reste null

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Methoxyphenol

Spécifications

CAS 150-76-5
Melting Point 55°C to 57°C (lit.)
Boiling Point 243°C (lit.)
Molecular Formula C7H8O2
Linear Formula CH3OC6H4OH
MDL Number MFCD00002332
Quantity 1 kg
Synonym p-Methoxyphenol; 4-Hydroxyanisole; 4-MP; HQMME; Hydroquinone monomethyl ether; MEHQ; MQ-F; p-Guaiacol
Molecular Weight (g/mol) 124.14
Percent Purity 99%