missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich 4-Methoxyphenol
Description
4-Methoxyphenol
Specifications
Specifications
| CAS | 150-76-5 |
| Melting Point | 55°C to 57°C (lit.) |
| Boiling Point | 243°C (lit.) |
| Molecular Formula | C7H8O2 |
| Linear Formula | CH3OC6H4OH |
| MDL Number | MFCD00002332 |
| Quantity | 100 g |
| Synonym | p-Methoxyphenol; 4-Hydroxyanisole; 4-MP; HQMME; Hydroquinone monomethyl ether; MEHQ; MQ-F; p-Guaiacol |
| Molecular Weight (g/mol) | 124.14 |
| Percent Purity | 99% |