missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxyphenethyl alcohol, 96%
CAS: 702-23-8 | C9H12O2 | 152.19 g/mol
$153.03 - $153.03
Chemical Identifiers
| CAS | 702-23-8 |
|---|---|
| Molecular Formula | C9H12O2 |
| Molecular Weight (g/mol) | 152.19 |
| MDL Number | MFCD00002900 |
| InChI Key | AUWDOZOUJWEPBA-UHFFFAOYSA-N |
| Synonym | 2-4-methoxyphenyl ethanol, 4-methoxyphenethyl alcohol, 4-methoxybenzeneethanol, benzeneethanol, 4-methoxy, p-methoxyphenethyl alcohol, 2-4-methoxyphenyl ethan-1-ol, unii-3mwl5nze9k, 3mwl5nze9k, 4-2-hydroxyethyl anisole, 2-4-methoxyphenyl-ethanol |
| PubChem CID | 69705 |
| IUPAC Name | 2-(4-methoxyphenyl)ethanol |
| SMILES | COC1=CC=C(C=C1)CCO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC354050100
|
Thermo Scientific Chemicals
354050100 |
10 g | Glass bottle |
Each for $153.03
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 702-23-8 | |
| 152.19 | |
| AUWDOZOUJWEPBA-UHFFFAOYSA-N | |
| 69705 | |
| COC1=CC=C(C=C1)CCO |
| C9H12O2 | |
| MFCD00002900 | |
| 2-4-methoxyphenyl ethanol, 4-methoxyphenethyl alcohol, 4-methoxybenzeneethanol, benzeneethanol, 4-methoxy, p-methoxyphenethyl alcohol, 2-4-methoxyphenyl ethan-1-ol, unii-3mwl5nze9k, 3mwl5nze9k, 4-2-hydroxyethyl anisole, 2-4-methoxyphenyl-ethanol | |
| 2-(4-methoxyphenyl)ethanol |
Specifications
| 702-23-8 | |
| 100.0 | |
| Brown | |
| >112°C | |
| 95% min. (GC) | |
| C9H12O2 | |
| CH3OC6H4CH2CH2OH | |
| 10 g | |
| 2-4-methoxyphenyl ethanol, 4-methoxyphenethyl alcohol, 4-methoxybenzeneethanol, benzeneethanol, 4-methoxy, p-methoxyphenethyl alcohol, 2-4-methoxyphenyl ethan-1-ol, unii-3mwl5nze9k, 3mwl5nze9k, 4-2-hydroxyethyl anisole, 2-4-methoxyphenyl-ethanol | |
| COC1=CC=C(C=C1)CCO | |
| 152.19 | |
| 152.19 | |
| Liquid After Melting |
| 95.0 | |
| 28°C to 30°C | |
| 334°C to 336°C | |
| Authentic | |
| Glass bottle | |
| 1.5360 to 1.538 | |
| MFCD00002900 | |
| 06, 906 | |
| AUWDOZOUJWEPBA-UHFFFAOYSA-N | |
| 2-(4-methoxyphenyl)ethanol | |
| 69705 | |
| 96% | |
| 4-Methoxyphenethyl alcohol, 96% |
Safety and Handling
EINECSNumber : 211-866-6
TSCA : TSCA
RUO – Research Use Only