missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Methoxyphenacyl Bromide 98.0+%, TCI America™
Supplier: TCI America M100425G
Specifications
| 4′-Methoxyphenacyl Bromide | |
| 71°C | |
| MFCD00000201 | |
| 3261 | |
| XQJAHBHCLXUGEP-UHFFFAOYSA-N | |
| 2-bromo-1-(4-methoxyphenyl)ethan-1-one | |
| 4965 | |
| ≥98.0% (GC) |
| 2632-13-5 | |
| C9H9BrO2 | |
| 25 g | |
| 2-bromo-4'-methoxyacetophenone, 2-bromo-1-4-methoxyphenyl ethanone, ethanone, 2-bromo-1-4-methoxyphenyl, p-methoxyphenacyl bromide, 4'-methoxyphenacyl bromide, 2-bromo-1-4-methoxyphenyl ethan-1-one, 4-methoxyphenacyl bromide, bromomethyl 4-methoxyphenyl ketone, alpha-bromo-4-methoxyacetophenone, 2-bromo-4-methoxyacetophenone | |
| COC1=CC=C(C=C1)C(=O)CBr | |
| 229.07 | |
| 229.07 | |
| Crystalline Powder |
Chemical Identifiers
| 2632-13-5 | |
| 229.07 | |
| XQJAHBHCLXUGEP-UHFFFAOYSA-N | |
| 4965 | |
| COC1=CC=C(C=C1)C(=O)CBr |
| C9H9BrO2 | |
| MFCD00000201 | |
| 2-bromo-4'-methoxyacetophenone, 2-bromo-1-4-methoxyphenyl ethanone, ethanone, 2-bromo-1-4-methoxyphenyl, p-methoxyphenacyl bromide, 4'-methoxyphenacyl bromide, 2-bromo-1-4-methoxyphenyl ethan-1-one, 4-methoxyphenacyl bromide, bromomethyl 4-methoxyphenyl ketone, alpha-bromo-4-methoxyacetophenone, 2-bromo-4-methoxyacetophenone | |
| 2-bromo-1-(4-methoxyphenyl)ethan-1-one |
Safety and Handling
TSCA : No