accessibility menu, dialog, popup

UserName

H3COOBr

CAS RN 2632-13-5

IUPAC Name: 2-bromo-1-(4-methoxyphenyl)ethan-1-one
Synonyms: 2-bromo-1-(4-methoxyphenyl)ethanone
Molecular Weight (g/mol): 229.07
Molecular Formula: C9H9BrO2
InChi Key: XQJAHBHCLXUGEP-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)CBr
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (2)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)