missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxynaphthalene-1-carboxylic acid, 98%

Catalog No. AAH6443203
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6443203 1 g
AAH6443206 5 g
AAH6443214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6443203 Supplier Thermo Scientific Chemicals Supplier No. H6443203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13041-62-8 | C12H10O3 | 202.209 g/mol

Used in the preparation of α-aryl/pyridinyl ethanolamines as selective β3 adrenergic receptor agonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the preparation of α-aryl/pyridinyl ethanolamines as selective β3 adrenergic receptor agonists.

Solubility
Soluble in DMSO and chloroform. Slightly soluble in water.

Notes
Store away from acids and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 13041-62-8
Molecular Formula C12H10O3
Molecular Weight (g/mol) 202.209
MDL Number MFCD02575372
InChI Key WRQHSQDGYYDRMX-UHFFFAOYSA-N
Synonym 4-methoxy-1-naphthoic acid, 4-methoxy-naphthalene-1-carboxylic acid, 1-naphthalenecarboxylic acid, 4-methoxy, 1-naphthalenecarboxylicacid, 4-methoxy, 4-methoxynaphthalenecarboxylic acid, ksc181m4d, 4-methoxy-1-naphthalenecarboxylic acid
PubChem CID 765380
IUPAC Name 4-methoxynaphthalene-1-carboxylic acid
SMILES COC1=CC=C(C2=CC=CC=C21)C(=O)O

Specifications

Melting Point 243°C to 247°C
Color White
Quantity 1 g
Beilstein 2099800
Solubility Information Soluble in DMSO and chloroform. Slightly soluble in water.
Formula Weight 202.21
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 4-Methoxynaphthalene-1-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only