missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxychalcone, 97%

Catalog No. AAL0331906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0331906 5 g
AAL0331914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0331906 Supplier Thermo Scientific Chemicals Supplier No. L0331906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 959-33-1 | C16H14O2 | 238.286 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 959-33-1
Molecular Formula C16H14O2
Molecular Weight (g/mol) 238.286
MDL Number MFCD00017179
InChI Key XUFXKBJMCRJATM-FMIVXFBMSA-N
Synonym 4-methoxychalcone, e-3-4-methoxyphenyl-1-phenylprop-2-en-1-one, 2e-3-4-methoxyphenyl-1-phenylprop-2-en-1-one, chalcone, 4-methoxy, 3-4-methoxyphenyl-1-phenyl-2-propen-1-one, 2-propen-1-one, 3-4-methoxyphenyl-1-phenyl, 2-propen-1-one, 3-4-methoxyphenyl-1-phenyl-, 2e, phenyl p-methoxystyryl ketone, 4'-methoxybenzylideneacetophenone, 3-4-methoxyphenyl-1-phenylprop-2-en-1-one
PubChem CID 641819
IUPAC Name (E)-3-(4-methoxyphenyl)-1-phenylprop-2-en-1-one
SMILES COC1=CC=C(C=C1)C=CC(=O)C2=CC=CC=C2

Specifications

Melting Point 73°C to 76°C
Quantity 5 g
Beilstein 978742
Formula Weight 238.29
Percent Purity 97%
Chemical Name or Material 4-Methoxychalcone
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-499-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only