missing translation for 'onlineSavingsMsg'
Learn More

(4-Methoxycarbonylbenzyl)triphenylphosphonium chloride, 97%

Catalog No. AAB2172903
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2172903 1 g
AAB2172906 5 g
AAB2172914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2172903 Supplier Thermo Scientific Chemicals Supplier No. B2172903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1253-47-0 | C27H24ClO2P | 446.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1253-47-0
Molecular Formula C27H24ClO2P
Molecular Weight (g/mol) 446.91
MDL Number MFCD02683069
InChI Key NKKAHXNONUTMEY-UHFFFAOYSA-M
Synonym 4-methoxycarbonylbenzyl triphenylphosphonium chloride, 4-methoxycarbonyl-benzyltriphenylphosphoniumchloride, 4-methoxycarbonyl benzyl triphenylphosphonium chloride, 4-carbomethoxybenzyl triphenylphosphonium chloride, methyl 4-triphenylphosphino methyl benzoate, chloride, acmc-1btgg, 4-methoxycarbonyl-benzyltriphenylphosphonium chloride, methyl alpha triphenylphosphonio-p-toluate chloride, 4-methoxycarbonyl-benzyl-triphenyl-phosphonium chloride
PubChem CID 18001058
IUPAC Name (4-methoxycarbonylphenyl)methyl-triphenylphosphanium;chloride
SMILES [Cl-].COC(=O)C1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1

Specifications

Melting Point 230°C to 235°C
Quantity 1 g
Sensitivity Hygroscopic
Formula Weight 446.91
Percent Purity 97%
Chemical Name or Material (4-Methoxycarbonylbenzyl)triphenylphosphonium chloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only