missing translation for 'onlineSavingsMsg'
Learn More

4-(Methoxycarbonyl)phenyl isothiocyanate, 98+%

Catalog No. AAL1047403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1047403 1 g
AAL1047406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1047403 Supplier Thermo Scientific Chemicals Supplier No. L1047403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3662-78-0 | C9H7NO2S | 193.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3662-78-0
Molecular Formula C9H7NO2S
Molecular Weight (g/mol) 193.22
MDL Number MFCD00041099
InChI Key WIZODHILGBTPPA-UHFFFAOYSA-N
Synonym 4-methoxycarbonylphenyl isothiocyanate, 4-methoxycarbonylphenylisothiocyanate, 4-isothiocyanatobenzoic acid methyl ester, benzoic acid, 4-isothiocyanato-, methyl ester, acmc-20amez, #, 4-methoxycarbonyl phenylisothiocyanate, 4-methoxycarbonyl phenyl isothiocyanate
PubChem CID 138002
IUPAC Name methyl 4-isothiocyanatobenzoate
SMILES COC(=O)C1=CC=C(C=C1)N=C=S

Specifications

Melting Point 46°C to 50°C
Odor Irritating
Quantity 1 g
UN Number UN2923
Beilstein 2100504
Sensitivity Moisture sensitive
Formula Weight 193.23
Percent Purity ≥98%
Chemical Name or Material 4-(Methoxycarbonyl)phenyl isothiocyanate
Hazard Category H302+H312-H314-H331-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only