missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxycarbonyl-2-nitrobenzeneboronic acid, 97%

Catalog No. AAH5286303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5286303 1 g
AAH5286306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5286303 Supplier Thermo Scientific Chemicals Supplier No. H5286303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 85107-55-7 | C8H8BNO6 | 224.963 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 85107-55-7
Molecular Formula C8H8BNO6
Molecular Weight (g/mol) 224.963
MDL Number MFCD01632202
InChI Key SFEJGHJCGQNFQC-UHFFFAOYSA-N
Synonym 4-methoxycarbonyl-2-nitrophenyl boronic acid, 4-methoxycarbonyl-2-nitrophenylboronic acid, 4-methoxycarbonyl-2-nitrobenzeneboronic acid, methyl 4-dihydroxyboranyl-3-nitrobenzoate, 2-nitro-4-methoxycarbonyl phenylboronic acid, 4-methoxycarbonyl-2-nitro-phenyl boronic acid, methyl 4-borono-3-nitrobenzoate, acmc-209q4d
PubChem CID 2773493
IUPAC Name (4-methoxycarbonyl-2-nitrophenyl)boronic acid
SMILES B(C1=C(C=C(C=C1)C(=O)OC)[N+](=O)[O-])(O)O

Specifications

Melting Point 160°C to 171°C
Quantity 1 g
Formula Weight 224.96
Percent Purity 97%
Chemical Name or Material 4-Methoxycarbonyl-2-nitrobenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only