missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybiphenyl, 98+%

CAS: 613-37-6 | C13H12O | 184.24 g/mol

$57.89 - $151.45

Chemical Identifiers

CAS 613-37-6
Molecular Formula C13H12O
Molecular Weight (g/mol) 184.24
MDL Number MFCD00014897
InChI Key RHDYQUZYHZWTCI-UHFFFAOYSA-N
Synonym 4-methoxybiphenyl, 4-phenylanisole, p-methoxybiphenyl, 1,1'-biphenyl, 4-methoxy, 4-methoxy-1,1'-biphenyl, anisole, p-phenyl, p-phenylanisole, 4-methoxy-biphenyl, biphenyl, 4-methoxy, 1-methoxy-4-phenyl-benzene
PubChem CID 11943
IUPAC Name 1-methoxy-4-phenylbenzene
SMILES COC1=CC=C(C=C1)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0126006
View Documents
Thermo Scientific Chemicals
L0126006
5 g N/A
Only null left
 
AAL0126014
View Documents
Thermo Scientific Chemicals
L0126014
25 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

613-37-6
184.24
RHDYQUZYHZWTCI-UHFFFAOYSA-N
11943
COC1=CC=C(C=C1)C1=CC=CC=C1
C13H12O
MFCD00014897
4-methoxybiphenyl, 4-phenylanisole, p-methoxybiphenyl, 1,1'-biphenyl, 4-methoxy, 4-methoxy-1,1'-biphenyl, anisole, p-phenyl, p-phenylanisole, 4-methoxy-biphenyl, biphenyl, 4-methoxy, 1-methoxy-4-phenyl-benzene
1-methoxy-4-phenylbenzene

Specifications

613-37-6
157°C (10 mmHg)
MFCD00014897
2043157
RHDYQUZYHZWTCI-UHFFFAOYSA-N
1-methoxy-4-phenylbenzene
11943
≥98%
86°C to 90°C
C13H12O
5 g
4-methoxybiphenyl, 4-phenylanisole, p-methoxybiphenyl, 1,1'-biphenyl, 4-methoxy, 4-methoxy-1,1'-biphenyl, anisole, p-phenyl, p-phenylanisole, 4-methoxy-biphenyl, biphenyl, 4-methoxy, 1-methoxy-4-phenyl-benzene
COC1=CC=C(C=C1)C1=CC=CC=C1
184.24
184.24
4-Methoxybiphenyl
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 210-339-8

RTECSNumber : BZ8850000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only