missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybiphenyl, 98+%

Catalog No. AAL0126006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0126006 5 g
AAL0126014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0126006 Supplier Thermo Scientific Chemicals Supplier No. L0126006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 613-37-6 | C13H12O | 184.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 613-37-6
Molecular Formula C13H12O
Molecular Weight (g/mol) 184.24
MDL Number MFCD00014897
InChI Key RHDYQUZYHZWTCI-UHFFFAOYSA-N
Synonym 4-methoxybiphenyl, 4-phenylanisole, p-methoxybiphenyl, 1,1'-biphenyl, 4-methoxy, 4-methoxy-1,1'-biphenyl, anisole, p-phenyl, p-phenylanisole, 4-methoxy-biphenyl, biphenyl, 4-methoxy, 1-methoxy-4-phenyl-benzene
PubChem CID 11943
IUPAC Name 1-methoxy-4-phenylbenzene
SMILES COC1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 86°C to 90°C
Boiling Point 157°C (10 mmHg)
Quantity 5 g
Beilstein 2043157
Formula Weight 184.24
Percent Purity ≥98%
Chemical Name or Material 4-Methoxybiphenyl
EINECSNumber 210-339-8
RTECSNumber BZ8850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only