missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzylamine 97.0+%, TCI America™

Catalog No. p-7132415
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M087025ML 25 mL
M0870250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M087025ML Supplier TCI America Supplier No. M087025ML
Only null left

Chemical Identifiers

CAS 2393-23-9
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00008122
InChI Key IDPURXSQCKYKIJ-UHFFFAOYSA-N
Synonym 4-methoxybenzylamine, 4-methoxyphenyl methanamine, benzenemethanamine, 4-methoxy, p-methoxybenzylamine, 4-methoxy-benzylamine, anisylamine, benzylamine, p-methoxy, 4-methoxybenzyl amine, 1-4-methoxyphenyl methanamine, 4-methoxyphenyl methylamine
PubChem CID 75452
ChEBI CHEBI:49837
IUPAC Name (4-methoxyphenyl)methanamine
SMILES COC1=CC=C(C=C1)CN

Specifications

Color Yellow
Boiling Point 236°C
Quantity 25 mL
UN Number 2735
Formula Weight 137.18
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Methoxybenzylamine