missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzylamine, 98%

CAS: 2393-23-9 | C8H11NO | 137.18 g/mol

$116.07 - $293.57

Chemical Identifiers

CAS 2393-23-9
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
InChI Key IDPURXSQCKYKIJ-UHFFFAOYSA-N
Synonym 4-methoxybenzylamine, 4-methoxyphenyl methanamine, benzenemethanamine, 4-methoxy, p-methoxybenzylamine, 4-methoxy-benzylamine, anisylamine, benzylamine, p-methoxy, 4-methoxybenzyl amine, 1-4-methoxyphenyl methanamine, 4-methoxyphenyl methylamine
PubChem CID 75452
ChEBI CHEBI:49837
IUPAC Name (4-methoxyphenyl)methanamine
SMILES COC1=CC=C(C=C1)CN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC125780250
SDS View Documents
Thermo Scientific Chemicals
125780250
25 mL Glass bottle
Each for $116.07
Only null left
 
AC125781000
SDS View Documents
Thermo Scientific Chemicals
125781000
100 mL Glass bottle
Each for $293.57
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2393-23-9
137.18
4-methoxybenzylamine, 4-methoxyphenyl methanamine, benzenemethanamine, 4-methoxy, p-methoxybenzylamine, 4-methoxy-benzylamine, anisylamine, benzylamine, p-methoxy, 4-methoxybenzyl amine, 1-4-methoxyphenyl methanamine, 4-methoxyphenyl methylamine
CHEBI:49837
COC1=CC=C(C=C1)CN
C8H11NO
IDPURXSQCKYKIJ-UHFFFAOYSA-N
75452
(4-methoxyphenyl)methanamine

Specifications

2393-23-9
1.0500g/mL
>110°C
97.5% min. (GC)
C8H11NO
CH3OC6H4CH2NH2
13,606
4-methoxybenzylamine, 4-methoxyphenyl methanamine, benzenemethanamine, 4-methoxy, p-methoxybenzylamine, 4-methoxy-benzylamine, anisylamine, benzylamine, p-methoxy, 4-methoxybenzyl amine, 1-4-methoxyphenyl methanamine, 4-methoxyphenyl methylamine
COC1=CC=C(C=C1)CN
137.18
CHEBI:49837
98%
4-Methoxybenzylamine
Colorless to Yellow
236.0°C to 237.0°C
Authentic
Glass bottle
1.5460 to 1.5480
25 mL
1.05
IDPURXSQCKYKIJ-UHFFFAOYSA-N
(4-methoxyphenyl)methanamine
75452
137.18
Liquid
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

EINECSNumber : 219-247-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only