missing translation for 'onlineSavingsMsg'
Learn More

(4-Methoxybenzyl)triphenylphosphonium chloride, 97%

Catalog No. AAL0059409
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0059409 10 g
AAL0059418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0059409 Supplier Thermo Scientific Chemicals Supplier No. L0059409

CAS: 3462-97-3 | C26H24ClOP | 418.90 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3462-97-3
Molecular Formula C26H24ClOP
Molecular Weight (g/mol) 418.90
MDL Number MFCD00031572
InChI Key YQXBNCFNXOFWLR-UHFFFAOYSA-M
Synonym 4-methoxybenzyl triphenylphosphonium chloride, 4-methoxyphenyl methyl triphenylphosphanium chloride, acmc-1ctv8, 4-methoxyphenyl methyl-triphenyl-phosphanium chloride, 4-methoxybenzyltriphenylphosphonium chloride, 4-methoxybenzyltriphenylphosphonium-chloride, 4-methoxybenzyl triphenyphosphonium chloride, p-methoxybenzyl-triphenylphosphonium chloride, 4-methoxybenzyltriphenyl phosphonium chloride, 4-methoxybenzyl triphenyl phosphonium chloride
PubChem CID 2773651
IUPAC Name (4-methoxyphenyl)methyl-triphenylphosphanium;chloride
SMILES [Cl-].COC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1

Specifications

Melting Point 235°C to 239°C
Quantity 10 g
Beilstein 4173378
Formula Weight 418.91
Percent Purity 97%
Chemical Name or Material (4-Methoxybenzyl)triphenylphosphonium chloride
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only