missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzyl Chloride (stabilized with Amylene) 98.0+%, TCI America™

Catalog No. M067625ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M067625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M067625ML Supplier TCI America Supplier No. M067625ML

Chemical Identifiers

CAS 824-94-2
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.609
MDL Number MFCD00000915
InChI Key MOHYOXXOKFQHDC-UHFFFAOYSA-N
Synonym 4-methoxybenzyl chloride, 4-methoxybenzylchloride, 1-chloromethyl-4-methoxybenzene, p-methoxybenzyl chloride, p-anisyl chloride, alpha-chloro-4-methoxytoluene, p-chloromethyl anisole, benzene, 1-chloromethyl-4-methoxy, 4-chloromethyl anisole, anisole, p-chloromethyl
PubChem CID 69993
IUPAC Name 1-(chloromethyl)-4-methoxybenzene
SMILES COC1=CC=C(C=C1)CCl

Specifications

Color Yellow
Boiling Point 127°C
Quantity 25 mL
UN Number 3265
Formula Weight 156.61
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Methoxybenzyl Chloride (stabilized with Amylene)
TSCA No
Recommended Storage Refrigerator