missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzyl 3-Chloromethyl-7-(2-phenylacetamido)-3-cephem-4-carboxylate 98.0+%, TCI America™

Catalog No. p-7133210
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24115G 5 g
M241125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24115G Supplier TCI America Supplier No. M24115G
Only null left

Chemical Identifiers

CAS 104146-10-3
Molecular Formula C24H23ClN2O5S
Molecular Weight (g/mol) 486.97
MDL Number MFCD00191253,MFCD00191253
InChI Key KFCMZNUGNLCSJQ-UHFFFAOYNA-N
Synonym 3-Chloromethyl-7-(2-phenylacetamido)-3-cephem-4-carboxylic Acid 4-Methoxybenzyl Ester
PubChem CID 13822075
IUPAC Name (4-methoxyphenyl)methyl 3-(chloromethyl)-8-oxo-7-(2-phenylacetamido)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMILES COC1=CC=C(COC(=O)C2=C(CCl)CSC3C(NC(=O)CC4=CC=CC=C4)C(=O)N23)C=C1

Specifications

Melting Point 153°C
Color Yellow
Quantity 5 g
Formula Weight 486.97
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxybenzyl 3-Chloromethyl-7-(2-phenylacetamido)-3-cephem-4-carboxylate
EINECSNumber (5)-6305
RTECSNumber XI0369883
TSCA No
Recommended Storage Freezer