accessibility menu, dialog, popup

UserName

CAS RN 104146-10-3

IUPAC Name: (4-methoxyphenyl)methyl 3-(chloromethyl)-8-oxo-7-(2-phenylacetamido)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Synonyms: (4-methoxyphenyl)methyl 3-(chloromethyl)-8-oxo-7-(2-phenylacetamido)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Molecular Weight (g/mol): 486.97
Molecular Formula: C24H23ClN2O5S
InChi Key: KFCMZNUGNLCSJQ-UHFFFAOYNA-N
SMILES: COC1=CC=C(COC(=O)C2=C(CCl)CSC3C(NC(=O)CC4=CC=CC=C4)C(=O)N23)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results