missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxybenzyl 3-Chloromethyl-7-(2-phenylacetamido)-3-cephem-4-carboxylate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 153°C |
| Color | Yellow |
| Quantity | 25 g |
| Formula Weight | 486.97 |
| Percent Purity | ≥98.0% (N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Methoxybenzyl 3-Chloromethyl-7-(2-phenylacetamido)-3-cephem-4-carboxylate |
Safety and Handling
| EINECSNumber | (5)-6305 |
| RTECSNumber | XI0369883 |
| TSCA | No |
| Recommended Storage | Freezer |