missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzyl 2,2,2-Trichloroacetimidate 96.0+%, TCI America™

Catalog No. M20165G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20165G 5 g
M201625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20165G Supplier TCI America Supplier No. M20165G
Only null left

Chemical Identifiers

CAS 89238-99-3
Molecular Formula C10H10Cl3NO2
Molecular Weight (g/mol) 282.545
MDL Number MFCD00134547
InChI Key TYHGKLBJBHACOI-UHFFFAOYSA-N
Synonym 2,2,2-Trichloroacetimidic Acid 4-Methoxybenzyl Ester
PubChem CID 11087263
IUPAC Name (4-methoxyphenyl)methyl 2,2,2-trichloroethanimidate
SMILES COC1=CC=C(C=C1)COC(=N)C(Cl)(Cl)Cl

Specifications

Color Yellow
Boiling Point 137°C
Quantity 5 g
Formula Weight 282.55
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methoxybenzyl 2,2,2-Trichloroacetimidate
TSCA No
Recommended Storage Freezer